Krios G4 Cryo-TEM powered by the Falcon 4 Direct Electron Detector and EPU Software*
Break down the barriers to cryo-EM adoption with the new Thermo Scientific Krios Solution; a suite of ease-of-use and throughput improvements as well as the most compact 300 kV cryo-EM microscope on the market. Scientists at leading research institutes and pharmaceutical companies around the globe already use our solutions to expand their understanding of disease and accelerate their drug discovery. The new Krios Solution can help you solve more structures faster than ever, making it possible to obtain high-resolution information of ever smaller proteins and biomolecules.
* All products are available separately in our Cryo-TEM portfolio.
3D reconstruction of the 20S proteasome from Thermoplasma acidophilum at 2.0 Angstrom resolution. Data collected on the Krios G4 Cryo-TEM with the Falcon 4 Direct Electron Detector. Images courtesy of Dr. Juergen Plitzko and Dr. Philipp Erdmann, Max Planck Institute for Biochemistry, Martinsried, Germany.
High productivity in the most compact design
The new Thermo Scientific Krios G4 Cryo-Transmission Electron Microscope (Cryo-TEM) is the most compact TEM in its class. A thorough redesign of the mechanical base frame and system enclosure reduces microscope height to below 3 meters. This allows the instrument to be installed in labs with a ceiling height below 3.04 m (~10 ft), avoiding costly room renovations.
The Krios G4 Cryo-TEM has improved ergonomics for easier sample exchange. Data acquisition set up is easier and faster thanks to enhanced automation, systematic user guidance and advanced performance monitoring. Simultaneously, the system’s productivity and high-resolution performance have been further enhanced through a combination of new optical modes, improved optical specifications and on-the-fly pre-processing.
Enhanced productivity
- Improved drift performance allows for a 3x faster experiment start.
- Powerful combination of fringe-free imaging (FFI), aberration-free image shift (AFIS) and the (optional) Thermo Scientific Falcon™ 4 Direct Electron Detector (with highest DQE in its class) enhances single particle analysis throughput by up to 400%.
Best-in-class image quality
Optical performance: 0.12 nm information limit and linear distortion of <0.5%
3D reconstruction of the 20S proteasome from Thermoplasma acidophilum at 2.0 Angstrom resolution. Data collected on the Krios G4 Cryo-TEM with the Falcon 4 Direct Electron Detector. Images courtesy of Dr. Juergen Plitzko and Dr. Philipp Erdmann, Max Planck Institute for Biochemistry, Martinsried, Germany.
Enhanced productivity, high throughput and improved image quality
With improved detector quantum efficiency (DQE) and a 10-times shorter exposure time over the entire spatial frequency range, the Falcon 4 Detector is truly the next generation in direct electron detection. The new optimized chip design is based on the proven large pixel layout of the Falcon product line (14 µm), further improving the unsurpassed low-noise levels and electron localization for reduced coincidence noise. Combined with an increased internal framerate (250 fps), best-in-class images can now be collected with a high throughput.
Additional features
- Next generation controller for minimized offload times
- Fully embedded in Thermo Scientific EPU and Tomography Software, guaranteeing smooth daily operation
- Built-in EPU Data Management (powered by Thermo Scientific Athena™ Software)
- Optional EPU Quality Monitor for on-the-fly quality assessment
Automated data acquisition software for Cryo-EM
Thermo Scientific EPU Software enables the automated collection of large numbers of vitrified particles from pre-selected areas on a grid and automates the operation of your instrument, ensuring optimal conditions before images are collected. New features include;
EPU Quality Monitor
This new addition improves productivity with uninterrupted on-the-fly pre-processing that lets you remotely monitor the quality of data as it’s being collected, allowing you to optimize data collection as it’s happening.
Data Management
Powered by Athena Software, this secured platform allows you to easily organize, visualize and share results of experimental workflows (including via remote access).